accessibility menu, dialog, popup

UserName

HOO

CAS RN 836-43-1

IUPAC Name: [4-(benzyloxy)phenyl]methanol
Synonyms: 4-benzyloxybenzyl alcohol
Molecular Weight (g/mol): 214.26
Molecular Formula: C14H14O2
InChi Key: OEBIVOHKFYSBPE-UHFFFAOYSA-N
SMILES: OCC1=CC=C(OCC2=CC=CC=C2)C=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)