CAS RN 832695-88-2
IUPAC Name:
[3-(methylcarbamoyl)phenyl]boronic acid
Synonyms:
3-(methylcarbamoyl)phenylboronic acid
Molecular Weight (g/mol):
178.98
Molecular Formula:
C8H10BNO3
InChi Key:
FYFFPNFUVMBPRZ-UHFFFAOYSA-N
SMILES:
CNC(=O)C1=CC=CC(=C1)B(O)O
1
–
1
of
1
results
1
Promotions Available
3-(Methylcarbamoyl)benzeneboronic acid, 98%
3-(Methylcarbamoyl)benzeneboronic acid, 98%
CAS: 832695-88-2 Molecular Formula: C8H10BNO3 Molecular Weight (g/mol): 178.98 MDL Number: MFCD04038918 InChI Key: FYFFPNFUVMBPRZ-UHFFFAOYSA-N Synonym: 3-n-methylaminocarbonyl phenylboronic acid,3-methylcarbamoyl phenyl boronic acid,3-methylcarbamoyl benzeneboronic acid,3-methylcarbamoyl phenylboronic acid,3-n-methylaminocarbonyl benzeneboronic acid,3-borono-n-methylbenzamide,boronic acid, 3-methylamino carbonyl phenyl,acmc-209prq,ksc496e8l PubChem CID: 2773524 IUPAC Name: [3-(methylcarbamoyl)phenyl]boronic acid SMILES: CNC(=O)C1=CC=CC(=C1)B(O)O