CAS RN 83265-53-6
IUPAC Name:
2-amino-5-(trifluoromethyl)benzoic acid
Synonyms:
2-amino-5-(trifluoromethyl)benzoic acid
Molecular Weight (g/mol):
205.14
Molecular Formula:
C8H6F3NO2
InChi Key:
GLCQUPLYYXSPQB-UHFFFAOYSA-N
SMILES:
NC1=C(C=C(C=C1)C(F)(F)F)C(O)=O
1
–
1
of
1
results
1
Promotions Available
2-Amino-5-(trifluoromethyl)benzoic acid, 95%
2-Amino-5-(trifluoromethyl)benzoic acid, 95%
CAS: 83265-53-6 Molecular Formula: C8H6F3NO2 Molecular Weight (g/mol): 205.136 MDL Number: MFCD01569537 InChI Key: GLCQUPLYYXSPQB-UHFFFAOYSA-N Synonym: 2-amino-5-trifluoromethyl benzoic acid,2-amino-5-trifluoromethyl-benzoic acid,5-trifluoromethylanthranilic acid,5-trifluoromethyl anthranilic acid,benzoic acid, 2-amino-5-trifluoromethyl,2-amino-5-trifluoromethyl-benzoicacid,pubchem12904,4-amino-3-carboxybenzotrifluoride,2-carboxy-4-trifluoromethyl aniline,2-amino-5-trifluoromethyl benzoicacid PubChem CID: 10465535 IUPAC Name: 2-amino-5-(trifluoromethyl)benzoic acid SMILES: C1=CC(=C(C=C1C(F)(F)F)C(=O)O)N