missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-(trifluoromethyl)benzoic acid, 95%

Catalog No. AAH64141MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH64141MD 250 mg
AAH6414103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH64141MD Supplier Thermo Scientific Chemicals Supplier No. H64141MD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 83265-53-6 | C8H6F3NO2 | 205.136 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 83265-53-6
Molecular Formula C8H6F3NO2
Molecular Weight (g/mol) 205.136
MDL Number MFCD01569537
InChI Key GLCQUPLYYXSPQB-UHFFFAOYSA-N
Synonym 2-amino-5-trifluoromethyl benzoic acid, 2-amino-5-trifluoromethyl-benzoic acid, 5-trifluoromethylanthranilic acid, 5-trifluoromethyl anthranilic acid, benzoic acid, 2-amino-5-trifluoromethyl, 2-amino-5-trifluoromethyl-benzoicacid, pubchem12904, 4-amino-3-carboxybenzotrifluoride, 2-carboxy-4-trifluoromethyl aniline, 2-amino-5-trifluoromethyl benzoicacid
PubChem CID 10465535
IUPAC Name 2-amino-5-(trifluoromethyl)benzoic acid
SMILES C1=CC(=C(C=C1C(F)(F)F)C(=O)O)N

Specifications

Melting Point 141°C to 146°C (decomposition)
Flash Point >110°C (230°F)
Quantity 250 mg
Formula Weight 205.14
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 2-Amino-5-(trifluoromethyl)benzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only