accessibility menu, dialog, popup

UserName

HNONOS

CAS RN 832115-62-5

IUPAC Name: N-cyclopropyl-5-(thiophen-2-yl)-1,2-oxazole-3-carboxamide
Synonyms: N-cyclopropyl-5-(thiophen-2-yl)-1,2-oxazole-3-carboxamide
Molecular Weight (g/mol): 234.27
Molecular Formula: C11H10N2O2S
InChi Key: SYENTKHGMVKGAQ-UHFFFAOYSA-N
SMILES: O=C(NC1CC1)C1=NOC(=C1)C1=CC=CS1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results