CAS RN 830-93-3
IUPAC Name:
[(5-bromo-1H-indol-3-yl)methyl]dimethylamine
Synonyms:
[(5-bromo-1H-indol-3-yl)methyl]dimethylamine
Molecular Weight (g/mol):
253.14
Molecular Formula:
C11H13BrN2
InChi Key:
FSERHDPEOFYMMK-UHFFFAOYSA-N
SMILES:
CN(C)CC1=CNC2=CC=C(Br)C=C12
1
–
1
of
1
results
1
Promotions Available
5-Bromogramine
5-Bromogramine
CAS: 830-93-3 Molecular Formula: C11H13BrN2 Molecular Weight (g/mol): 253.14 InChI Key: FSERHDPEOFYMMK-UHFFFAOYSA-N Synonym: 5-bromogramine,1-5-bromo-1h-indol-3-yl-n,n-dimethylmethanamine,indole, 5-bromo-3-dimethylamino methyl,5-bromo-1h-indol-3-yl methyl dimethylamine,5-bromo-3-dimethylaminomethyl indole,5-bromogramine crystalline,5-bromo-n,n-dimethyl-1h-indole-3-methanamine,1h-indole-3-methanamine, 5-bromo-n,n-dimethyl,4-22-00-04316 beilstein handbook reference,5-bromo-1h-indol-3-ylmethyl-dimethyl-amine PubChem CID: 13249 IUPAC Name: 1-(5-bromo-1H-indol-3-yl)-N,N-dimethylmethanamine SMILES: CN(C)CC1=CNC2=C1C=C(C=C2)Br