accessibility menu, dialog, popup

UserName

NH2SOOFFF

CAS RN 830-43-3

IUPAC Name: 4-(trifluoromethyl)benzene-1-sulfonamide
Synonyms: 4-(trifluoromethyl)benzenesulfonamide
Molecular Weight (g/mol): 225.19
Molecular Formula: C7H6F3NO2S
InChi Key: TVHXQQJDMHKGGK-UHFFFAOYSA-N
SMILES: NS(=O)(=O)C1=CC=C(C=C1)C(F)(F)F
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (2)
  • (2)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (2)
  • (1)
  • (3)