accessibility menu, dialog, popup

UserName

OHPHOOO(S)(R)OPHOOHO(r)OPHOOHO(S)OPHOHOO(R)OPOHHOO(s)OPOHHOO

CAS RN 83-86-3

IUPAC Name: {[(1R,2R,3S,4R,5s,6S)-2,3,4,5,6-pentakis(phosphonooxy)cyclohexyl]oxy}phosphonic acid
Synonyms: [(1R,2R,3S,4R,5s,6S)-2,3,4,5,6-pentakis(phosphonooxy)cyclohexyl]oxyphosphonic acid myo-inositol hexakisphosphate myo-inosistol hexakisphosphate hexaphosphate, inositol alkovert saeure des phytins phytin preparation inosithexaphosphorsaure hexakisphosphate, inositol sodium phytate (substance)
Molecular Weight (g/mol): 660.03
Molecular Formula: C6H18O24P6
InChi Key: IMQLKJBTEOYOSI-GPIVLXJGSA-N
SMILES: OP(O)(=O)O[C@H]1[C@H](OP(O)(O)=O)[C@@H](OP(O)(O)=O)[C@H](OP(O)(O)=O)[C@H](OP(O)(O)=O)[C@@H]1OP(O)(O)=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)