missing translation for 'onlineSavingsMsg'
Learn More

Phytic Acid (ca. 50% in Water, ca. 1.1mol/L), TCI America™

Catalog No. P0409500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P0409500G 500 g
P040925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P0409500G Supplier TCI America Supplier No. P0409500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 83-86-3
Molecular Formula C6H18O24P6
Molecular Weight (g/mol) 660.03
MDL Number MFCD00082309
InChI Key IMQLKJBTEOYOSI-GPIVLXJGSA-N
Synonym phytic acid, phytate, fytic acid, inositol hexaphosphate, alkalovert, myo-inositol hexaphosphate, alkovert, phytine, acide fytique, acidum fyticum
PubChem CID 890
IUPAC Name {[(1R,2R,3S,4R,5s,6S)-2,3,4,5,6-pentakis(phosphonooxy)cyclohexyl]oxy}phosphonic acid
SMILES OP(O)(=O)O[C@H]1[C@H](OP(O)(O)=O)[C@@H](OP(O)(O)=O)[C@H](OP(O)(O)=O)[C@H](OP(O)(O)=O)[C@@H]1OP(O)(O)=O

Specifications

Color Undesignated
Formula Weight 660.03
Quantity 500 g
Physical Form Clear Liquid at 20°C
Chemical Name or Material Phytic Acid (ca. 50% in Water, ca. 1.1mol/L)
EINECSNumber (3)-2516
RTECSNumber NM7525000
TSCA Yes
Recommended Storage Refrigerator