accessibility menu, dialog, popup

UserName

CAS RN 83-43-2

IUPAC Name: (1R,3aS,3bS,5S,9aR,9bS,10S,11aS)-1,10-dihydroxy-1-(2-hydroxyacetyl)-5,9a,11a-trimethyl-1H,2H,3H,3aH,3bH,4H,5H,7H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-7-one
Synonyms: methylprednisolone solomet besonia methylprednisolone preparation metastab suprametil methylprednisolone preparation (product) urbason promacortine methylprednisolone (substance)
Molecular Weight (g/mol): 374.48
Molecular Formula: C22H30O5
InChi Key: VHRSUDSXCMQTMA-PJHHCJLFSA-N
SMILES: C[C@H]1C[C@H]2[C@@H]3CC[C@](O)(C(=O)CO)[C@@]3(C)C[C@H](O)[C@@H]2[C@@]2(C)C=CC(=O)C=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results