missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 6-α-Methylprednisolone

Catalog No. AC338800010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC338800010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC338800010 Supplier Thermo Scientific Chemicals Supplier No. 338800010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 83-43-2
Molecular Formula C22H30O5
Molecular Weight (g/mol) 374.48
MDL Number MFCD00010591
InChI Key VHRSUDSXCMQTMA-PJHHCJLFSA-N
Synonym methylprednisolone, medrol, medrone, urbason, methylprednisolon, 6alpha-methylprednisolone, metilbetasone, medrate, metilprednisolone, dopomedrol
PubChem CID 6741
ChEBI CHEBI:6888
IUPAC Name (6S,8S,9S,10R,11S,13S,14S,17R)-11,17-dihydroxy-17-(2-hydroxyacetyl)-6,10,13-trimethyl-7,8,9,11,12,14,15,16-octahydro-6H-cyclopenta[a]phenanthren-3-one
SMILES C[C@H]1C[C@H]2[C@@H]3CC[C@](O)(C(=O)CO)[C@@]3(C)C[C@H](O)[C@@H]2[C@@]2(C)C=CC(=O)C=C12

Specifications

Melting Point 227.0°C to 231.0°C
Quantity 1 g
Merck Index 15,6184
Solubility Information Solubility in water: insoluble. Other solubilities: slightly soluble in acetone
Formula Weight 374.48
Chemical Name or Material 6-α-Methylprednisolone
Hazard Category GHS Signal Word: Danger
EINECSNumber 201-476-4

RUO – Research Use Only