accessibility menu, dialog, popup

UserName

OOONOO

CAS RN 82911-69-1

IUPAC Name: 2,5-dioxopyrrolidin-1-yl (9H-fluoren-9-yl)methyl carbonate
Synonyms: 2,5-dioxopyrrolidin-1-yl 9H-fluoren-9-ylmethyl carbonate
Molecular Weight (g/mol): 337.33
Molecular Formula: C19H15NO5
InChi Key: WMSUFWLPZLCIHP-UHFFFAOYSA-N
SMILES: O=C(OCC1C2=CC=CC=C2C2=CC=CC=C12)ON1C(=O)CCC1=O
Molecular Weight (g/mol) 
  • (2)
  • (5)
Percent Purity 
  • (2)
  • (5)
Quantity 
  • (1)
  • (3)
  • (3)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (2)
  • (5)
  • (2)
  • (5)
  • (1)
  • (3)
  • (3)
  • (2)
  • (2)