missing translation for 'onlineSavingsMsg'
Learn More

N-[(9H-Fluoren-9-ylmethoxy)carbonyloxy]succinimide 98.0+%, TCI America™

Catalog No. F02395G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F02395G 5 g
F023925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F02395G Supplier TCI America Supplier No. F02395G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 82911-69-1
Molecular Formula C19H15NO5
Molecular Weight (g/mol) 337.331
MDL Number MFCD00010733
InChI Key WMSUFWLPZLCIHP-UHFFFAOYSA-N
Synonym fmoc-osu, n-9-fluorenylmethoxycarbonyloxy succinimide, fmoc-onsu, 9-fluorenylmethyl succinimidyl carbonate, fmoc n-hydroxysuccinimide ester, 9h-fluoren-9-yl methyl 2,5-dioxopyrrolidin-1-yl carbonate, 9-fluorenylmethyl n-succinimidyl carbonate, 9-fluorenylmethylsuccinimidyl carbonate, 9-fluorenylmethoxycarbonyl-n-hydroxysuccinimide
PubChem CID 134122
IUPAC Name (2,5-dioxopyrrolidin-1-yl) 9H-fluoren-9-ylmethyl carbonate
SMILES C1CC(=O)N(C1=O)OC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24

Specifications

Melting Point 153°C
Color White
Quantity 5 g
Formula Weight 337.33
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-[(9H-Fluoren-9-ylmethoxy)carbonyloxy]succinimide
TSCA No
Recommended Storage Refrigerator