missing translation for 'onlineSavingsMsg'
Learn More

N-(9-Fluorenylmethoxycarbonyloxy)succinimide, 98%

Catalog No. AAA1314314
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1314314 25 g
AAA1314303 1 g
AAA1314306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1314314 Supplier Thermo Scientific Chemicals Supplier No. A1314314
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 82911-69-1 | C19H15NO5 | 337.331 g/mol

N-(9-Fluorenylmethoxycarbonyloxy)succinimide is the most efficient reagent for the selective preparation of N-(9-fluorenylmethoxycarbonyl) derivatives of hydroxy-amino acids. Dipeptide formation is lower than with the chloroformate, Fmoc-Cl. It is also employed in the synthesis of glycopeptides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-(9-Fluorenylmethoxycarbonyloxy)succinimide is the most efficient reagent for the selective preparation of N-(9-fluorenylmethoxycarbonyl) derivatives of hydroxy-amino acids. Dipeptide formation is lower than with the chloroformate, Fmoc-Cl. It is also employed in the synthesis of glycopeptides.

Solubility
Soluble in dimethyl sulfoxide and dimethyl formamide.

Notes
Store in a cool place. Moisture sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong acids.

Chemical Identifiers

CAS 82911-69-1
Molecular Formula C19H15NO5
Molecular Weight (g/mol) 337.331
MDL Number MFCD00010733
InChI Key WMSUFWLPZLCIHP-UHFFFAOYSA-N
Synonym fmoc-osu, n-9-fluorenylmethoxycarbonyloxy succinimide, fmoc-onsu, 9-fluorenylmethyl succinimidyl carbonate, fmoc n-hydroxysuccinimide ester, 9h-fluoren-9-yl methyl 2,5-dioxopyrrolidin-1-yl carbonate, 9-fluorenylmethyl n-succinimidyl carbonate, 9-fluorenylmethylsuccinimidyl carbonate, 9-fluorenylmethoxycarbonyl-n-hydroxysuccinimide
PubChem CID 134122
IUPAC Name (2,5-dioxopyrrolidin-1-yl) 9H-fluoren-9-ylmethyl carbonate
SMILES C1CC(=O)N(C1=O)OC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24

Specifications

Melting Point 150°C to 153°C
Quantity 25 g
UN Number UN3077
Beilstein 3569540
Sensitivity Moisture sensitive
Solubility Information Soluble in dimethyl sulfoxide and dimethyl formamide.
Formula Weight 337.34
Percent Purity 98%
Chemical Name or Material N-(9-Fluorenylmethoxycarbonyloxy)succinimide
Hazard Category H302-H317
Hazard Statement GHS H Statement
H301-H317
Toxic if swallowed.
May cause an allergic skin reaction.
Precautionary Statement P261-P264b-P270-P272-P280g-P301+P312-P302+P352-P330-P333+P313-P363-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber 433-520-5
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only