accessibility menu, dialog, popup

UserName

CH3H3CCH3OOHNOOH

CAS RN 82732-07-8

IUPAC Name: 2-{[(tert-butoxy)carbonyl]amino}-4-phenylbutanoic acid
Synonyms: 2-[(tert-butoxycarbonyl)amino]-4-phenylbutanoic acid
Molecular Weight (g/mol): 279.34
Molecular Formula: C15H21NO4
InChi Key: MCODLPJUFHPVQP-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)NC(CCC1=CC=CC=C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)