CAS RN 82702-31-6
IUPAC Name:
methyl 3-bromo-4-fluorobenzoate
Synonyms:
methyl 3-bromo-4-fluorobenzoate
Molecular Weight (g/mol):
233.04
Molecular Formula:
C8H6BrFO2
InChi Key:
JVORYGNKYAXATM-UHFFFAOYSA-N
SMILES:
COC(=O)C1=CC=C(F)C(Br)=C1
1
–
1
of
1
results
1
Methyl 3-Bromo-4-fluorobenzoate 98.0+%, TCI America™
CAS: 82702-31-6 Molecular Formula: C8H6BrFO2 Molecular Weight (g/mol): 233.04 MDL Number: MFCD04972915 InChI Key: JVORYGNKYAXATM-UHFFFAOYSA-N Synonym: 3-Bromo-4-fluorobenzoic Acid Methyl Ester PubChem CID: 7018022 IUPAC Name: methyl 3-bromo-4-fluorobenzoate SMILES: COC(=O)C1=CC=C(F)C(Br)=C1