accessibility menu, dialog, popup

UserName

OOFClNCl

CAS RN 82671-06-5

IUPAC Name: 2,6-dichloro-5-fluoropyridine-3-carboxylate
Synonyms: 2,6-dichloro-5-fluoropyridine-3-carboxylate
Molecular Weight (g/mol): 208.98
Molecular Formula: C6HCl2FNO2
InChi Key: LTDGKGCHRNNCAC-UHFFFAOYSA-M
SMILES: [O-]C(=O)C1=CC(F)=C(Cl)N=C1Cl
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results