accessibility menu, dialog, popup

UserName

CH3SiH3CCH3OPCl

CAS RN 82495-75-8

IUPAC Name: triphenyl({[2-(trimethylsilyl)ethoxy]methyl})phosphanium chloride
Synonyms: triphenyl({[2-(trimethylsilyl)ethoxy]methyl})phosphanium chloride
Molecular Weight (g/mol): 429.01
Molecular Formula: C24H30ClOPSi
InChi Key: NEUMNYXEDIPGJD-UHFFFAOYSA-M
SMILES: [Cl-].C[Si](C)(C)CCOC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results