accessibility menu, dialog, popup

UserName

CAS RN 82419-26-9

IUPAC Name: 2,3-difluoro-6-nitrobenzen-1-olate
Synonyms: 2,3-difluoro-6-nitrobenzenolate
Molecular Weight (g/mol): 174.08
Molecular Formula: C6H2F2NO3
InChi Key: KEGOHDCHURMFKX-UHFFFAOYSA-M
SMILES: [O-]C1=C(F)C(F)=CC=C1[N+]([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results