CAS RN 824-69-1
IUPAC Name:
2,5-dichlorobenzene-1,4-diol
Synonyms:
2,5-dichlorohydroquinone 2,5-dichloro-1,4-hydroquinone 2,5-dichloro-p-hydroquinone
Molecular Weight (g/mol):
179
Molecular Formula:
C6H4Cl2O2
InChi Key:
AYNPIRVEWMUJDE-UHFFFAOYSA-N
SMILES:
OC1=CC(Cl)=C(O)C=C1Cl
1
–
1
of
1
results
1
Promotions Available
2,5-Dichlorohydroquinone, 97%
2,5-Dichlorohydroquinone, 97%
CAS: 824-69-1 Molecular Formula: C6H4Cl2O2 Molecular Weight (g/mol): 179.00 MDL Number: MFCD00041749 InChI Key: AYNPIRVEWMUJDE-UHFFFAOYSA-N Synonym: 2,5-dichlorohydroquinone,1,4-benzenediol, 2,5-dichloro,2,5-dichloro-p-hydroquinone,hydroquinone, 2,5-dichloro,2,5-dichloro-p-benzohydroquinone,2,5-dchq,2,5-dichloro-1,4-hydroquinone,2,5-dichloro-1,4-benzenediol,unii-d489x4tqvd,2,5-dichloro-1,4-dihydroxybenzene PubChem CID: 65 ChEBI: CHEBI:27545 IUPAC Name: 2,5-dichlorobenzene-1,4-diol SMILES: OC1=CC(Cl)=C(O)C=C1Cl