accessibility menu, dialog, popup

UserName

H3CO

CAS RN 823-76-7

IUPAC Name: 1-cyclohexylethan-1-one
Synonyms: ethanone, 1-cyclohexyl-
Molecular Weight (g/mol): 126.2
Molecular Formula: C8H14O
InChi Key: RIFKADJTWUGDOV-UHFFFAOYSA-N
SMILES: CC(=O)C1CCCCC1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)