CAS RN 823-57-4
CAS RN 823-57-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2-Bromo-5-chloroaniline 97.0+%, TCI America™
CAS: 823-57-4 Molecular Formula: C6H5BrClN Molecular Weight (g/mol): 206.467 MDL Number: MFCD00672942 InChI Key: NLEZSQHAFMZAGU-UHFFFAOYSA-N PubChem CID: 11310276 IUPAC Name: 2-bromo-5-chloroaniline SMILES: C1=CC(=C(C=C1Cl)N)Br