accessibility menu, dialog, popup

UserName

CAS RN 82113-65-3

IUPAC Name: 1,1,1-trifluoro-N-trifluoromethanesulfonylmethanesulfonamide
Synonyms: 1,1,1-trifluoro-N-trifluoromethanesulfonylmethanesulfonamide
Molecular Weight (g/mol): 281.14
Molecular Formula: C2HF6NO4S2
InChi Key: ZXMGHDIOOHOAAE-UHFFFAOYSA-N
SMILES: FC(F)(F)S(=O)(=O)NS(=O)(=O)C(F)(F)F
Boiling Point 
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (3)
Physical Form 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)