accessibility menu, dialog, popup

UserName

Enjoy Up to 50% Off Spotlight Deals All Year Long

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Main Menu
Documents and Certificates

CAS RN 819050-89-0

CH3H3CCH3H3COOOOCH3H3CCH3H3COOOORhRh

CAS RN 819050-89-0

IUPAC Name: bis(λ²-rhodium(2+)) bis(3-[3-(2-carboxylato-2,2-dimethylethyl)phenyl]-2,2-dimethylpropanoate)
Synonyms: bis(λ²-rhodium(2+)) bis(3-[3-(2-carboxylato-2,2-dimethylethyl)phenyl]-2,2-dimethylpropanoate)
Molecular Weight (g/mol): 758.48
Molecular Formula: C32H40O8Rh2
InChi Key: AHCPGXZDVSBJII-UHFFFAOYSA-J
SMILES: [Rh++].[Rh++].CC(C)(CC1=CC(CC(C)(C)C([O-])=O)=CC=C1)C([O-])=O.CC(C)(CC1=CC(CC(C)(C)C([O-])=O)=CC=C1)C([O-])=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.