accessibility menu, dialog, popup

UserName

CAS RN 818-44-0

IUPAC Name: ethenyl octanoate
Synonyms: octanoic acid, vinyl ester (mixed isomers)
Molecular Weight (g/mol): 170.25
Molecular Formula: C10H18O2
InChi Key: QBDADGJLZNIRFQ-UHFFFAOYSA-N
SMILES: CCCCCCCC(=O)OC=C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results