accessibility menu, dialog, popup

UserName

H3CSCl

CAS RN 817181-75-2

IUPAC Name: 2-chloro-3-hexylthiophene
Synonyms: 2-chloro-3-hexylthiophene
Molecular Weight (g/mol): 202.74
Molecular Formula: C10H15ClS
InChi Key: BSWSVVYUCAKNDK-UHFFFAOYSA-N
SMILES: CCCCCCC1=C(Cl)SC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results