CAS RN 81536-18-7
IUPAC Name:
2-(2-methylpropane-2-sulfonyl)acetonitrile
Synonyms:
2-(2-methylpropane-2-sulfonyl)acetonitrile
Molecular Weight (g/mol):
161.22
Molecular Formula:
C6H11NO2S
InChi Key:
ZRCYCIRRPIYJOU-UHFFFAOYSA-N
SMILES:
CC(C)(C)S(=O)(=O)CC#N
1
–
1
of
1
results
1
Promotions Available
tert-Butylsulfonylacetonitrile, 98%
tert-Butylsulfonylacetonitrile, 98%
CAS: 81536-18-7 Molecular Formula: C6H11NO2S Molecular Weight (g/mol): 161.219 MDL Number: MFCD00051622 InChI Key: ZRCYCIRRPIYJOU-UHFFFAOYSA-N Synonym: tert-butylsulfonylacetonitrile,2-tert-butylsulfonyl acetonitrile,2-2-methylpropane-2-sulfonyl acetonitrile,2-tert-butylsulfonylethanenitrile,2-tert-butyl sulfonyl ethanenitrile,acmc-1ailq,maybridge1_004678,t-butylsulfonylacetonitrile,tert-butanesulfonylacetonitrile,tert-butylsulfonyl acetonitrile PubChem CID: 2778536 IUPAC Name: 2-tert-butylsulfonylacetonitrile SMILES: CC(C)(C)S(=O)(=O)CC#N