CAS RN 815-85-0
IUPAC Name:
λ²-tin(2+) 2,3-dihydroxybutanedioate
Synonyms:
tin tartrate
Molecular Weight (g/mol):
266.78
Molecular Formula:
C4H4O6Sn
InChi Key:
YXTDAZMTQFUZHK-UHFFFAOYNA-L
SMILES:
[Sn++].OC(C(O)C([O-])=O)C([O-])=O
1
–
1
of
1
results
1
Tin(II) tartrate hydrate, 95%
CAS: 815-85-0 Molecular Formula: C4H4O6Sn Molecular Weight (g/mol): 266.78 MDL Number: MFCD00054358 InChI Key: YXTDAZMTQFUZHK-UHFFFAOYNA-L Synonym: unii-6js9u5611x,stannous tartrate,tin tartrate,tartaric acid, tin 2+ salt 1:1,r-r*,r*-2,3-dihydroxybutanedioic acid,tin r-r*,r*-tartrate,butanedioic acid, 2,3-dihydroxy-, r-r*,r*-, tin 2+ salt 1:1,butanedioic acid, 2,3-dihydroxy-theta-theta,theta-, tin 2++ salt 1:1 PubChem CID: 66545835 IUPAC Name: (2R,3R)-2,3-dihydroxybutanedioate;tin(2+) SMILES: [Sn++].OC(C(O)C([O-])=O)C([O-])=O