accessibility menu, dialog, popup

UserName

FFBrFFFFFFFFBrFFFFFF

CAS RN 812-58-8

IUPAC Name: 1,8-dibromo-1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8-hexadecafluorooctane
Synonyms: 1,8-dibromo-1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8-hexadecafluorooctane
Molecular Weight (g/mol): 559.87
Molecular Formula: C8Br2F16
InChi Key: LYRJPHOHVRHNQH-UHFFFAOYSA-N
SMILES: FC(F)(Br)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)Br
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results