accessibility menu, dialog, popup

UserName

CAS RN 81167-39-7

H3COON

CAS RN 81167-39-7

IUPAC Name: methyl 2-[(diphenylmethylidene)amino]acetate
Synonyms: methyl 2-[(diphenylmethylidene)amino]acetate
Molecular Weight (g/mol): 253.3
Molecular Formula: C16H15NO2
InChi Key: PQTOLHHWLUCKSB-UHFFFAOYSA-N
SMILES: COC(=O)CN=C(C1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)