accessibility menu, dialog, popup

UserName

CAS RN 811-93-8

CH3CH3H2NH2N

CAS RN 811-93-8

IUPAC Name: 2-methylpropane-1,2-diamine
Synonyms: 1,2-propanediamine, 2-methyl-
Molecular Weight (g/mol): 88.15
Molecular Formula: C4H12N2
InChi Key: OPCJOXGBLDJWRM-UHFFFAOYSA-N
SMILES: CC(C)(N)CN
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results