accessibility menu, dialog, popup

UserName

CAS RN 81015-49-8

IUPAC Name: 4-(2,3-dihydro-1,3-thiazol-2-ylidene)cyclohexa-2,5-dien-1-one
Synonyms: 4-(3H-1,3-thiazol-2-ylidene)cyclohexa-2,5-dien-1-one
Molecular Weight (g/mol): 177.22
Molecular Formula: C9H7NOS
InChi Key: LMMPCMRUGFBCPV-UHFFFAOYSA-N
SMILES: O=C1C=CC(C=C1)=C1NC=CS1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results