accessibility menu, dialog, popup

UserName

HHClClNH2HNHO

CAS RN 81012-99-9

IUPAC Name: dihydrogen 3-(hydrazinylmethyl)phenol dichloride
Synonyms: dihydrogen 3-(hydrazinylmethyl)phenol dichloride
Molecular Weight (g/mol): 211.09
Molecular Formula: C7H12Cl2N2O
InChi Key: ONOJPUDFIOEGCX-UHFFFAOYSA-N
SMILES: [H+].[H+].[Cl-].[Cl-].NNCC1=CC=CC(O)=C1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results