accessibility menu, dialog, popup

UserName

CAS RN 81-21-0

IUPAC Name: 4,10-dioxapentacyclo[6.3.1.0²,⁷.0³,⁵.0⁹,¹¹]dodecane
Synonyms: dicyclopentadiene dioxide
Molecular Weight (g/mol): 164.2
Molecular Formula: C10H12O2
InChi Key: BQQUFAMSJAKLNB-UHFFFAOYNA-N
SMILES: C1C2OC2C2C3CC(C4OC34)C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results