accessibility menu, dialog, popup

UserName

CAS RN 81-11-8

IUPAC Name: 5-amino-2-[(1E)-2-(4-amino-2-sulfophenyl)ethenyl]benzene-1-sulfonic acid
Synonyms: amsonic acid
Molecular Weight (g/mol): 370.39
Molecular Formula: C14H14N2O6S2
InChi Key: REJHVSOVQBJEBF-OWOJBTEDSA-N
SMILES: NC1=CC=C(\C=C\C2=CC=C(N)C=C2S(O)(=O)=O)C(=C1)S(O)(=O)=O
Molecular Weight (g/mol) 
  • (5)
  • (2)
Percent Purity 
  • (3)
  • (4)
Quantity 
  • (3)
  • (2)
  • (2)
Physical Form 
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (3)
  • (4)
  • (5)
  • (2)
  • (3)
  • (4)
  • (3)
  • (2)
  • (2)
  • (5)