accessibility menu, dialog, popup

UserName

CH3OOHNH2HCl

CAS RN 80997-87-1

IUPAC Name: 2-amino-3-phenylbutanoic acid hydrochloride
Synonyms: 2-amino-3-phenylbutanoic acid hydrochloride
Molecular Weight (g/mol): 215.68
Molecular Formula: C10H14ClNO2
InChi Key: SGKWQBMDOXGYAA-UHFFFAOYNA-N
SMILES: Cl.CC(C(N)C(O)=O)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)