accessibility menu, dialog, popup

UserName

CAS RN 80822-15-7

IUPAC Name: (2S,3S)-2,3-bis(benzoyloxy)butanedioic acid hydrate
Synonyms: (2S,3S)-2,3-bis(benzoyloxy)butanedioic acid hydrate
Molecular Weight (g/mol): 376.32
Molecular Formula: C18H16O9
InChi Key: DXDIHODZARUBLA-DZWQFRRJNA-N
SMILES: O.OC(=O)[C@@H](OC(=O)C1=CC=CC=C1)[C@H](OC(=O)C1=CC=CC=C1)C(O)=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results