accessibility menu, dialog, popup

UserName

CAS RN 80806-68-4

IUPAC Name: 4,4,5,5,6,6,7,7,8,8,9,9,9-tridecafluorononan-1-ol
Synonyms: 4,4,5,5,6,6,7,7,8,8,9,9,9-tridecafluorononan-1-ol
Molecular Weight (g/mol): 378.13
Molecular Formula: C9H7F13O
InChi Key: HMGDEQANNRNNKX-UHFFFAOYSA-N
SMILES: OCCCC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results