accessibility menu, dialog, popup

UserName

CH3OSiCH3H3COSiCH3CH3H3COSiO

CAS RN 8050-81-5

IUPAC Name: 3,3,5,5-tetramethyl-2,4-dioxa-3,5-disilahexane; silanedione
Synonyms: simethicone
Molecular Weight (g/mol): 238.46
Molecular Formula: C6H18O4Si3
InChi Key: AMTWCFIAVKBGOD-UHFFFAOYSA-N
SMILES: O=[Si]=O.CO[Si](C)(C)O[Si](C)(C)C
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results