accessibility menu, dialog, popup

UserName

ZnCuAl109

CAS RN 8049-11-4

IUPAC Name: nonaaluminium decacopper zinc
Synonyms: nonaaluminium decacopper zinc
Molecular Weight (g/mol): 943.67
Molecular Formula: Al9Cu10Zn
InChi Key: MPKKOQSNGWAMJD-UHFFFAOYSA-N
SMILES: [Al].[Al].[Al].[Al].[Al].[Al].[Al].[Al].[Al].[Cu].[Cu].[Cu].[Cu].[Cu].[Cu].[Cu].[Cu].[Cu].[Cu].[Zn]
Form 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)