accessibility menu, dialog, popup

UserName

NCH3H2NNH2NNH2H2NCl

CAS RN 8048-52-0

IUPAC Name: 3,6-diamino-10-methylacridin-10-ium acridine-3,6-diamine chloride
Synonyms: acriflavine
Molecular Weight (g/mol): 468.99
Molecular Formula: C27H25ClN6
InChi Key: PEJLNXHANOHNSU-UHFFFAOYSA-N
SMILES: [Cl-].NC1=CC2=NC3=CC(N)=CC=C3C=C2C=C1.C[N+]1=C2C=C(N)C=CC2=CC2=CC=C(N)C=C12
Physical Form 
  • (1)
  • (1)
  • (2)
Quantity 
  • (2)
  • (2)
Percent Purity 
  • (1)
  • (2)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)