accessibility menu, dialog, popup

UserName

CAS RN 80466-78-0

IUPAC Name: 3,5-dimethyl-1H-pyrazole-4-sulfonyl chloride
Synonyms: 3,5-dimethyl-1H-pyrazole-4-sulfonyl chloride
Molecular Weight (g/mol): 194.63
Molecular Formula: C5H7ClN2O2S
InChi Key: NBGSZCQLPLYKJH-UHFFFAOYSA-N
SMILES: CC1=C(C(C)=NN1)S(Cl)(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results