accessibility menu, dialog, popup

UserName

CAS RN 80366-85-4

H3CCH3H3COOHNOOONOO

CAS RN 80366-85-4

IUPAC Name: 2,5-dioxopyrrolidin-1-yl 2-({[(tert-butoxy)carbonyl]amino}oxy)acetate
Synonyms: 2,5-dioxopyrrolidin-1-yl 2-{[(tert-butoxycarbonyl)amino]oxy}acetate
Molecular Weight (g/mol): 288.26
Molecular Formula: C11H16N2O7
InChi Key: ZZBOTLREHORFCK-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)NOCC(=O)ON1C(=O)CCC1=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results