CAS RN 8032-32-4
Petroleum Ether (Certified ACS), Fisher Chemical™
CAS: 8032-32-4 MDL Number: MFCD00081849 Synonym: Ligroine, Benzine, Naphtha Petroleum, Naphtha Solvent (Optima/Pesticide/Certified ACS),magnesium, bromo 4-methylphenyl,4-methylphenylmagnesium bromide,bromo 4-methylphenyl magnesium,4-tolylmagnesium bromide,4-toluylmagnesium bromide,p-tolyl-magnesium bromide,4-tolyl magnesium bromide,4-methylphenylmagnesiumbromide,p-methylphenylmagnesium bromide
Petroleum Ether, Optima™ for HPLC and GC, Fisher Chemical™
MDL Number: MFCD00081849
Petroleum Ether (Pesticide), Fisher Chemical™
CAS: 8032-32-4 MDL Number: MFCD00081849 Synonym: Ligroine,Benzine,Naphtha Petroleum,Naphtha Solvent,Petroleum Benzine,4-toluylmagnesium bromide,p-tolyl-magnesium bromide,4-tolyl magnesium bromide,4-methylphenylmagnesiumbromide,p-methylphenylmagnesium bromide
Petroleum ether 60/80
CAS: 8032-32-4 MDL Number: MFCD00081849 Synonym: p-tolylmagnesium bromide,magnesium, bromo 4-methylphenyl,4-methylphenylmagnesium bromide,bromo 4-methylphenyl magnesium,4-tolylmagnesium bromide,4-toluylmagnesium bromide,p-tolyl-magnesium bromide,4-tolyl magnesium bromide,4-methylphenylmagnesiumbromide,p-methylphenylmagnesium bromide
Petroleum Ether, 35° to 60°C, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™
CAS: 8032-32-4 Synonym: p-tolylmagnesium bromide,magnesium, bromo 4-methylphenyl,4-methylphenylmagnesium bromide,bromo 4-methylphenyl magnesium,4-tolylmagnesium bromide,4-toluylmagnesium bromide,p-tolyl-magnesium bromide,4-tolyl magnesium bromide,4-methylphenylmagnesiumbromide,p-methylphenylmagnesium bromide
Petrol, puriss., 100° to 140°C, Solstice
CAS: 8032-32-4 MDL Number: MFCD00081849 Synonym: p-tolylmagnesium bromide,magnesium, bromo 4-methylphenyl,4-methylphenylmagnesium bromide,bromo 4-methylphenyl magnesium,4-tolylmagnesium bromide,4-toluylmagnesium bromide,p-tolyl-magnesium bromide,4-tolyl magnesium bromide,4-methylphenylmagnesiumbromide,p-methylphenylmagnesium bromide
Petroleum ether 35/60, ACS
CAS: 8032-32-4 MDL Number: MFCD00081849 Synonym: p-tolylmagnesium bromide,magnesium, bromo 4-methylphenyl,4-methylphenylmagnesium bromide,bromo 4-methylphenyl magnesium,4-tolylmagnesium bromide,4-toluylmagnesium bromide,p-tolyl-magnesium bromide,4-tolyl magnesium bromide,4-methylphenylmagnesiumbromide,p-methylphenylmagnesium bromide
Petroleum Ether, Spectrophotometric Grade, Solstice
CAS: 8032-32-4 MDL Number: MFCD00081849 Synonym: p-tolylmagnesium bromide,magnesium, bromo 4-methylphenyl,4-methylphenylmagnesium bromide,bromo 4-methylphenyl magnesium,4-tolylmagnesium bromide,4-toluylmagnesium bromide,p-tolyl-magnesium bromide,4-tolyl magnesium bromide,4-methylphenylmagnesiumbromide,p-methylphenylmagnesium bromide
Petroleum Ether, ACS Reagent, for organic synthesis, prep-LC, and general laboratory use, >99.0% (C5 and C6 Isomers), Solstice
CAS: 8032-32-4 Synonym: p-tolylmagnesium bromide,magnesium, bromo 4-methylphenyl,4-methylphenylmagnesium bromide,bromo 4-methylphenyl magnesium,4-tolylmagnesium bromide,4-toluylmagnesium bromide,p-tolyl-magnesium bromide,4-tolyl magnesium bromide,4-methylphenylmagnesiumbromide,p-methylphenylmagnesium bromide
Petroleum ether 40/60
CAS: 8032-32-4 MDL Number: MFCD00081849 Synonym: p-tolylmagnesium bromide,magnesium, bromo 4-methylphenyl,4-methylphenylmagnesium bromide,bromo 4-methylphenyl magnesium,4-tolylmagnesium bromide,4-toluylmagnesium bromide,p-tolyl-magnesium bromide,4-tolyl magnesium bromide,4-methylphenylmagnesiumbromide,p-methylphenylmagnesium bromide
Petroleum ether 35/60, HPLC Grade
CAS: 8032-32-4 MDL Number: MFCD00081849 Synonym: p-tolylmagnesium bromide,magnesium, bromo 4-methylphenyl,4-methylphenylmagnesium bromide,bromo 4-methylphenyl magnesium,4-tolylmagnesium bromide,4-toluylmagnesium bromide,p-tolyl-magnesium bromide,4-tolyl magnesium bromide,4-methylphenylmagnesiumbromide,p-methylphenylmagnesium bromide
Petroleum ether 35/60, Environmental Grade
CAS: 8032-32-4 MDL Number: MFCD00081849 Synonym: p-tolylmagnesium bromide,magnesium, bromo 4-methylphenyl,4-methylphenylmagnesium bromide,bromo 4-methylphenyl magnesium,4-tolylmagnesium bromide,4-toluylmagnesium bromide,p-tolyl-magnesium bromide,4-tolyl magnesium bromide,4-methylphenylmagnesiumbromide,p-methylphenylmagnesium bromide
Petroleum Ether, Macron Fine Chemicals™
CAS: 8032-32-4 Synonym: p-tolylmagnesium bromide,magnesium, bromo 4-methylphenyl,4-methylphenylmagnesium bromide,bromo 4-methylphenyl magnesium,4-tolylmagnesium bromide,4-toluylmagnesium bromide,p-tolyl-magnesium bromide,4-tolyl magnesium bromide,4-methylphenylmagnesiumbromide,p-methylphenylmagnesium bromide
Petroleum Ether Analytical Grade, Lab Chem
CAS: 8032-32-4 Molecular Weight (g/mol): 90
Petroleum Ether, ACS Reagent (Ligroin), Reagents
Petroleum Ether, ACS Reagent (Ligroin), ACS, for general laboratory use. Manufactured in ISO 9001 facility.
- 1
- 2