missing translation for 'onlineSavingsMsg'
Learn More

Petroleum ether 35/60, ACS

Catalog No. AA42085M6
Change view
Click to view available options
Quantity:
1 L
4 L
20 L
4 x 1 L
4 x 4 L
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA42085M6 4 x 4 L
AA42085K2 1 L
AA42085M1 4 x 1 L
AA42085K7 4 L
AA42085M4 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. AA42085M6 Supplier Thermo Scientific Chemicals Supplier No. 042085M6

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Maximum level of impurities: Color (APHA): 10, Boiling range 35-60°C, Evaporation residue: 0.001%, Acidity P.T. | CAS: 8032-32-4

Petroleum ether is mostly used by pharmaceutical companies and in the manufacturing process. It is a lightweight hydrocarbon used chiefly as a nonpolar solvent. The extract of aerial parts of ageratum conyzoids in petroleum ether plays an essential role for the isolation of sigmasterol and beta sitosterol. Crude petroleum spirit and methanol extracts also play an important role for the screening of antimicrobial activity of the stem bark, leaves and roots of ficus thoningii.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 8032-32-4
MDL Number MFCD00081849
Synonym p-tolylmagnesium bromide, magnesium, bromo 4-methylphenyl, 4-methylphenylmagnesium bromide, bromo 4-methylphenyl magnesium, 4-tolylmagnesium bromide, 4-toluylmagnesium bromide, p-tolyl-magnesium bromide, 4-tolyl magnesium bromide, 4-methylphenylmagnesiumbromide, p-methylphenylmagnesium bromide

Specifications

Color Colorless
Appearance Colorless liquid
Density 0.64
Quantity 4 x 4 L
Flash Point -49°C (-57°F)
Ignition Point 250°C (482°F)
Odor Petroleum-like
Physical Form Liquid
Refractive Index 1.363
Vapor Pressure 350 hPa (263mm Hg) at 20°C
Boiling Point 35°C to 60°C
Solubility Information Insoluble in water.
Grade ACS
Chemical Name or Material Petroleum ether 35/60
Show More Show Less
Hazard Category H225-H304-H315-H319-H336
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P312-P331-P332+P313-P363-P370+P378q-P501c
DOTInformation Hazard Class: 3; Packaging Group: II
EINECSNumber 232-453-7
RTECSNumber OI6180000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only