accessibility menu, dialog, popup

UserName

CAS RN 80241-45-8

IUPAC Name: [(pyridin-2-yl)methyl]phosphonic acid
Synonyms: pyridin-2-ylmethylphosphonic acid
Molecular Weight (g/mol): 173.11
Molecular Formula: C6H8NO3P
InChi Key: WVWCWWLEFNGDBC-UHFFFAOYSA-N
SMILES: OP(O)(=O)CC1=CC=CC=N1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results