accessibility menu, dialog, popup

UserName

CAS RN 80041-89-0

H3CCH3BOHHO

CAS RN 80041-89-0

IUPAC Name: (propan-2-yl)boronic acid
Synonyms: isopropylboronic acid
Molecular Weight (g/mol): 87.91
Molecular Formula: C3H9BO2
InChi Key: QIPHSSYCQCBJAX-UHFFFAOYSA-N
SMILES: CC(C)B(O)O
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results