missing translation for 'onlineSavingsMsg'
Learn More

Benzalkonium Chloride, MilliporeSigma™ Supelco™

Catalog No. 111002556
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111002556 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
Catalog No. 111002556 missing translation for 'mfr' Merck Emd Millipore missing translation for 'supplierNo' 10509935ML
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia. Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 8001-54-5
Purity Grade Notes These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.
Type USP Reference Standard
Quantity 5 mL
Grade Pharmaceutical Primary Standard