missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzalkonium Chloride, 95-104%, Spectrum™ Chemical
Description
Spectrum™ Chemical Benzalkonium Chloride is used as a biocide, a phase transfer agent, and a cationic surfactant in the chemical industry. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.
Specifications
Specifications
| CAS Max % | 100% |
| Physical Form | Adhering Powder and/or Lumps |
| Quantity | 100 g |
| UN Number | UN2923 |
| Percent Purity | 95 to 104% |
| Grade | Ungraded |
| Packaging | Glass Bottle |
| Chemical Name or Material | Benzalkonium Chloride |