accessibility menu, dialog, popup

UserName

CH3OOCH3H2C

CAS RN 80-62-6

IUPAC Name: methyl 2-methylprop-2-enoate
Synonyms: methyl methacrylate zimmer bone cement simplex P monomer, methylmethacrylate methyl methylmethacrylate methyl monomer sintex methyl-methacrylat bone cement, zimmer MMA cranioplast
Molecular Weight (g/mol): 100.12
Molecular Formula: C5H8O2
InChi Key: VVQNEPGJFQJSBK-UHFFFAOYSA-N
SMILES: COC(=O)C(C)=C
Percent Purity 
  • (2)
  • (6)
  • (6)
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)
  • (3)
Boiling Point 
  • (2)
  • (3)
  • (3)
Molecular Weight (g/mol) 
  • (5)
  • (3)
Grade 
  • (6)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (12)
  • (6)
  • (6)
  • (2)
  • (6)
  • (2)
  • (6)
  • (6)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)
  • (3)
  • (2)
  • (3)
  • (3)
  • (5)
  • (3)
  • (6)
  • (3)
  • (2)